5-Bromo-5-nitro-1,3-dioxane ≥98.0%
Supplier: TCI America
Synonyms:
Bromonitrodioxane
, Bronidox L
B3156-25G
TCB3156-25GEA
148.99
USD
TCB3156-25G
5-Bromo-5-nitro-1,3-dioxane ≥98.0%
5-Bromo-5-nitro-1,3-dioxane
CAS Number: 30007-47-7
MDL Number: MFCD00101855
Molecular Formula: C4H6BrNO4
Molecular Weight: 212.00
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 61
Storage Temperature: 0-10°C
Formula:
C₄H₆BrNO₄ MW: 212.00 g/mol Melting Pt: 61 °C |
MDL Number:
MFCD00101855 CAS Number: 30007-47-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...