Alizarin cyanin green F

Supplier: TCI America

Synonyms: D and C Green No. 5Acid Green 25

A0610-25G
TCA0610-025GEA 80.39 USD
TCA0610-025G
Alizarin cyanin green F
Alizarin cyanin green F

CAS Number: 4403-90-1
MDL Number: MFCD00001193
Molecular Formula: C28H22N2O8S2
Molecular Weight: 622.57
Form: Crystal
Color: Deep Green
Melting point (°C): 238
Lambda max.: 641 nm (H2O)


Other Product Names / Synonyms: Acid Green 25

Formula: C₂₈H₂₂N₂O₈S₂
MW: 622.57 g/mol
Melting Pt: 238 °C
MDL Number: MFCD00001193
CAS Number: 4403-90-1
https://us.vwr.com/stibo/web/std.lang.all/80/29/28928029.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR