5-Bromo-2-tert-butylpyrimidine ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
B3079-1G
TCB3079-1GEA
354.57
USD
TCB3079-1G
5-Bromo-2-tert-butylpyrimidine ≥98.0% (by GC, total nitrogen)
5-Bromo-2-tert-butylpyrimidine
CAS Number: 85929-94-8
Molecular Formula: C8H11BrN2
Molecular Weight: 215.09
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 90
Melting point (°C): 53
Storage Temperature:
Formula:
C₈H₁₁BrN₂ Boiling Pt: 90 °C Melting Pt: 53 °C |
CAS Number:
85929-94-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...