Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride ≥98.0%

Supplier: TCI America

Synonyms: 3a,4,7,7a-Tetrahydro-4,7-ethanoisobenzofuran-1,3-dione

B3038-5G
TCB3038-5GEA 230.41 USD
TCB3038-5G
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride ≥98.0%
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride

CAS Number: 6708-37-8
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 148

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 148 °C
MDL Number: MFCD06796361
CAS Number: 6708-37-8
https://us.vwr.com/stibo/web/std.lang.all/44/81/28934481.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR