6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B3034-5G
B3034-1G
TCB3034-5GEA
436
USD
TCB3034-5G
TCB3034-1G
6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine ≥98.0% (by HPLC, titration analysis)
6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine
CAS Number: 92276-38-5
Molecular Formula: C5H3BrN4
Molecular Weight: 199.01
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 206
Other Product Names / Synonyms: 6-Bromo-4-azabenzotriazole
Formula:
C₅H₃BrN₄ Melting Pt: 206 °C |
MDL Number:
MFCD04060716 CAS Number: 92276-38-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...