Bis(4-nitrophenyl)amine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4'-Dinitrodiphenylamine

B2960-1G B2960-5G
TCB2960-5GEA 237.43 USD
TCB2960-5G TCB2960-1G
Bis(4-nitrophenyl)amine ≥98.0% (by HPLC)
Bis(4-nitrophenyl)amine

CAS Number: 1821-27-8
MDL Number: MFCD00437044
Molecular Formula: C12H9N3O4
Molecular Weight: 259.22
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 216


Other Product Names / Synonyms: 4,4'-Dinitrodiphenylamine

Formula: C₁₂H₉N₃O₄
MW: 259.22 g/mol
Melting Pt: 216 °C
MDL Number: MFCD00437044
CAS Number: 1821-27-8
https://us.vwr.com/stibo/web/std.lang.all/44/20/28934420.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR