2-(1,2-Benzisoxazol-3-yl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1,2-Benzisoxazole-3-acetic Acid
B2912-25G
TCB2912-25GEA
47.69
USD
TCB2912-25G
2-(1,2-Benzisoxazol-3-yl)acetic acid ≥98.0% (by GC, titration analysis)
2-(1,2-Benzisoxazol-3-yl)acetic acid
CAS Number: 4865-84-3
MDL Number: MFCD02180388
Molecular Formula: C9H7NO3
Molecular Weight: 177.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 129
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...