(S)-4-Benzyloxy-1,3-butanediol ≥96.0%

B2902-1G
TCB2902-1GEA 434.52 USD
TCB2902-1G
(S)-4-Benzyloxy-1,3-butanediol ≥96.0%
(S)-4-Benzyloxy-1,3-butanediol

CAS Number: 85418-23-1
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Storage Temperature: 0-10°C

Formula: C₁₁H₁₆O₃
Boiling Pt: 141 °C
MDL Number: MFCD11846118
CAS Number: 85418-23-1

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR