1-(2-Bromoisovaleryl)urea ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Bromovalerylurea
, 2-Bromo-N-carbamoyl-3-methylbutanamide
B2842-25G
TCB2842-25GEA
121
USD
TCB2842-25G
1-(2-Bromoisovaleryl)urea ≥98.0% (by titrimetric analysis)
1-(2-Bromoisovaleryl)urea
CAS Number: 496-67-3
Molecular Formula: C6H11BrN2O2
Molecular Weight: 223.07
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 152
Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction)
Other Product Names / Synonyms: Bromovalerylurea
Formula:
C₆H₁₁BrN₂O₂ MW: 223.07 g/mol Melting Pt: 152 °C |
MDL Number:
MFCD00047873 CAS Number: 496-67-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...