(1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane ≥95.0%

Supplier: TCI America

B2779-100MG
TCB2779-100MGEA 145.08 USD
TCB2779-100MG
(1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane ≥95.0%
(1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane

CAS Number: 387834-41-5
Molecular Formula: C19H39BrO2Si2
Molecular Weight: 435.59
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid

Formula: C₁₉H₃₉BrO₂Si₂
CAS Number: 387834-41-5
https://us.vwr.com/stibo/web/std.lang.all/42/95/28934295.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR