Mordant blue 13
Supplier: TCI America
Synonyms:
3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt
, Eriochrome Blue SE
, Sunchromine Fast Blue MB
, Plasmocorinth B disodium salt
, 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalenedisulphonic Acid Disodium Salt
, Plasmocorinth B
CAS Number: 1058-92-0
MDL Number: MFCD00003941
Molecular Formula: C16H11ClN2O9S2
Molecular Weight: 518.80
Form: Crystal
Color: Deep Yellow
Other Product Names / Synonyms: 3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, Eriochrome Blue SE, Sunchromine Fast Blue MB, Plasmocorinth B
Formula:
C₁₆H₁₁ClN₂O₉S₂ MW: 518.80 g/mol |
MDL Number:
MFCD00003941 CAS Number: 1058-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...