Azocal A
Supplier: TCI America
A0582-25G
TCA0582-025GEA
320
USD
TCA0582-025G
Azocal A
Azocal A
[for Calcium determination]
CAS Number: 1836-22-2
MDL Number: MFCD00046379
Molecular Formula: C17H12N2O9S2
Molecular Weight: 518.35
Form: Crystal
Color: Yellow Red
Formula:
C₁₇H₁₂N₂O₉S₂ |
MDL Number:
MFCD00046379 CAS Number: 1836-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...