9,9-Bis(4-amino-3-fluorophenyl)fluorene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
4,4'-(9H-Fluorene-9,9-diyl)bis(2-fluoroaniline)
, 2,2'-Difluoro-4,4'-(9-fluorenylidene)dianiline
B2691-5G
TCB2691-5GEA
127.13
USD
TCB2691-5G
9,9-Bis(4-amino-3-fluorophenyl)fluorene ≥98.0% (by HPLC, total nitrogen)
9,9-Bis(4-amino-3-fluorophenyl)fluorene
CAS Number: 127926-65-2
Molecular Formula: C25H18F2N2
Molecular Weight: 384.43
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2,2'-Difluoro-4,4'-(9-fluorenylidene)dianiline
Formula:
C₂₅H₁₈F₂N₂ MW: 384.43 g/mol |
MDL Number:
MFCD08276304 CAS Number: 127926-65-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...