9,9-Bis(4-amino-3-chlorophenyl)fluorene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2,2'-Dichloro-4,4'-(9-fluorenylidene)dianiline
B2690-5G
TCB2690-5GEA
211.03
USD
TCB2690-5G
9,9-Bis(4-amino-3-chlorophenyl)fluorene ≥98.0% (by HPLC, titration analysis)
9,9-Bis(4-amino-3-chlorophenyl)fluorene
CAS Number: 107934-68-9
Molecular Formula: C25H18Cl2N2
Molecular Weight: 417.33
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 200
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2,2'-Dichloro-4,4'-(9-fluorenylidene)dianiline
Formula:
C₂₅H₁₈Cl₂N₂ MW: 417.33 g/mol Melting Pt: 200 °C |
MDL Number:
MFCD10000948 CAS Number: 107934-68-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...