(2-Bromo-3-pyridinyl)methanol ≥98.0%
Supplier: TCI America
Synonyms:
2-Bromo-3-(hydroxymethyl)pyridine
, 2-Bromo-3-pyridinemethanol
, (2-Bromopyridin-3-yl)methanol
B2674-1G
TCB2674-1GEA
257.73
USD
TCB2674-1G
(2-Bromo-3-pyridinyl)methanol ≥98.0%
(2-Bromo-3-pyridinyl)methanol
CAS Number: 131747-54-1
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 75
Other Product Names / Synonyms: 2-Bromo-3-(hydroxymethyl)pyridine, 2-Bromonicotinyl Alcohol
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...