Alizarine yellow R ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Alizarin Yellow R5-(4-Nitrophenylazo)salicylic acidMordant Orange 1

A0579-25G
TCA0579-025GEA 76.13 USD
TCA0579-025G
Alizarine yellow R ≥98.0% (by HPLC)
Alizarine Yellow R

CAS Number: 2243-76-7
MDL Number: MFCD00007313
Molecular Formula: C13H9N3O5
Molecular Weight: 309.21
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Stains & Dyes, Analytical Science, Indicators (pH)
Other Product Names / Synonyms: Alizarin Yellow R, 5-(4-Nitrophenylazo)salicylic Acid

Formula: C₁₃H₉N₃O₅
MDL Number: MFCD00007313
CAS Number: 2243-76-7
EINECS: 218-818-3
https://us.vwr.com/stibo/web/std.lang.all/80/02/28928002.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR