9,9-Bis(4-aminophenyl)fluorene ≥99.0% (by HPLC, titration analysis), purified by sublimation

Supplier: TCI America

Synonyms: 4,4'-(9H-Fluorene-9,9-diyl)dianiline4,4'-(9-Fluorenylidene)dianiline

B2654-1G
TCB2654-1GEA 92.37 USD
TCB2654-1G
9,9-Bis(4-aminophenyl)fluorene ≥99.0% (by HPLC, titration analysis), purified by sublimation
9,9-Bis(4-aminophenyl)fluorene

CAS Number: 15499-84-0
MDL Number: MFCD00039156
Molecular Formula: C25H20N2
Molecular Weight: 348.45
Purity/Analysis Method: 99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 237


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents, Monomers, Polyimide Raw Materials, Highly Purified Reagents
Other Product Names / Synonyms: 4,4'-(9-Fluorenylidene)dianiline

Formula: C₂₅H₂₀N₂
MW: 348.45 g/mol
Melting Pt: 237 °C
MDL Number: MFCD00039156
CAS Number: 15499-84-0

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR