Alizarine yellow GG (sodium salt)
Supplier: TCI America
Synonyms:
Sodium 2-hydroxy-5-((3-nitrophenyl)diazenyl)benzoate
A0578-25G
TCA0578-025GEA
46.69
USD
TCA0578-025G
Alizarine yellow GG (sodium salt)
Alizarine Yellow GG (sodium salt)
CAS Number: 584-42-9
MDL Number: MFCD00044746
Molecular Formula: C13H9N3O5
Molecular Weight: 309.21
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: Metachrome Yellow, Salicyl Yellow
Formula:
C₁₃H₈N₃NaO₅ MW: 309.21 g/mol |
MDL Number:
MFCD00044746 CAS Number: 584-42-9 EINECS: 209-536-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...