Bis[3-(trimethoxysilyl)propyl]amine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bis(3-(trimethoxysilyl)propyl)amine

B2548-25G
TCB2548-25GEA 30.43 USD
TCB2548-25G
Bis[3-(trimethoxysilyl)propyl]amine ≥98.0% (by titrimetric analysis)
Bis[3-(trimethoxysilyl)propyl]amine

CAS Number: 82985-35-1
MDL Number: MFCD00191835
Molecular Formula: C12H31NO6Si2
Molecular Weight: 341.55
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 110
Specific Gravity (20/20): 1.05

Formula: C₁₂H₃₁NO₆Si₂
MW: 341.55 g/mol
Boiling Pt: 152 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00191835
CAS Number: 82985-35-1
https://us.vwr.com/stibo/web/std.lang.all/41/85/28934185.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR