(4aS,8aR)-2-Benzoyloctahydro-6(2H)isoquinolinone ≥90.0%

Supplier: TCI America

B2420-50MG
TCB2420-50MGEA 854 USD
TCB2420-50MG
(4aS,8aR)-2-Benzoyloctahydro-6(2H)isoquinolinone ≥90.0%
(4aS,8aR)-2-Benzoyloctahydro-6(2H)isoquinolinone

CAS Number: 52390-26-8
Molecular Formula: C16H19NO2
Molecular Weight: 257.33
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Specific rotation [a]20/D: -57 deg (C=1, MeOH)

Formula: C₁₆H₁₉NO₂
CAS Number: 52390-26-8
https://us.vwr.com/stibo/web/std.lang.all/41/51/28934151.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR