(1S,2S)-N,N'-Bis[(R)-2-hydroxy-2'-phenyl-1,1'-binaphthyl-3-ylmethylene]-1,2-diphenylethylenediaminato Manganese(III) Acetate ≥89.0% (by titrimetric analysis)
B2409-100MG
TCB2409-100MGEA
348.14
USD
TCB2409-100MG
(1S,2S)-N,N'-Bis[(R)-2-hydroxy-2'-phenyl-1,1'-binaphthyl-3-ylmethylene]-1,2-diphenylethylenediaminato Manganese(III) Acetate ≥89.0% (by titrimetric analysis)
(1S,2S)-N,N'-Bis[(R)-2-hydroxy-2'-phenyl-1,1'-binaphthyl-3-ylmethylene]-1,2-diphenylethylenediaminato manganese(III) acetate
CAS Number: 158052-18-7
MDL Number: MFCD06797076
Molecular Formula: C70H49MnN2O4
Molecular Weight: 1037.11
Purity/Analysis Method:
Form: Crystal
Formula:
C₇₀H₄₉MnN₂O₄ |
MDL Number:
MFCD06797076 CAS Number: 158052-18-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...