Ethyl-cis-3-bromoacrylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: cis-3-Bromoacrylic Acid Ethyl Ester

B2362-1G
TCB2362-1GEA 124 USD
TCB2362-1G
Ethyl-cis-3-bromoacrylate ≥98.0% (by GC, titration analysis)
Ethyl-cis-3-bromoacrylate

CAS Number: 31930-34-4
MDL Number: MFCD00674501
Molecular Formula: C5H7BrO2
Molecular Weight: 179.01
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 67
Specific Gravity (20/20): 1.48
Storage Temperature: 0-10°C


Other Product Names / Synonyms: cis-3-Bromoacrylic Acid Ethyl Ester

Formula: C₅H₇BrO₂
MW: 179.01 g/mol
Boiling Pt: 67 °C
Density: 1.48 (20 °C)
MDL Number: MFCD00674501
CAS Number: 31930-34-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR