TCIAB2340

B2340-1G
TCB2340-1GEA 53.43 USD
TCB2340-1G
TCIAB2340
CAS Number: 2264-01-9
MDL Number: MFCD00078412
Molecular Formula: C12H10F8O4
Molecular Weight: 370.20
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.44

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,2,3,3,4,4,5,5-Octafluorohexamethylene Diacrylate, 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol Diacrylate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR