(2-Bromopropyl)benzene ≥93.0%

Supplier: TCI America

Synonyms: 2-Bromo-1-phenylpropane(2-Bromopropyl)benzene

B2290-25G B2290-5G
TCB2290-5GEA 168.49 USD
TCB2290-5G TCB2290-25G
(2-Bromopropyl)benzene ≥93.0%
(2-Bromopropyl)benzene

CAS Number: 2114-39-8
MDL Number: MFCD00000149
Molecular Formula: C9H11Br
Molecular Weight: 199.09
Purity/Analysis Method: 93.0% (GC)
Form: Clear Liquid
Flash Point (°C): 90
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: (2-Bromopropyl)benzene

Formula: C₆H₅CH₂CHBrCH₃
MW: 199.09 g/mol
Density: 1.30 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00000149
CAS Number: 2114-39-8
https://us.vwr.com/stibo/web/std.lang.all/40/56/28934056.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR