8-Azaadenine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3H-[1,2,3]Triazolo[4,5-d]pyrimidin-7-amine

A0552-1G
TCA0552-001GEA 186.31 USD
TCA0552-001G
8-Azaadenine ≥98.0% (by HPLC)
8-Azaadenine

CAS Number: 1123-54-2
MDL Number: MFCD00005697
Molecular Formula: C4H4N6
Molecular Weight: 136.12
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 320

Formula: C₄H₄N₆
MW: 136.12 g/mol
Melting Pt: 320 °C
MDL Number: MFCD00005697
CAS Number: 1123-54-2
https://us.vwr.com/stibo/web/std.lang.all/79/82/28927982.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR