Bis(pentafluorophenyl)dimethylsilane ≥97.0%

B2277-1G
TCB2277-1GEA 67.32 USD
TCB2277-1G
Bis(pentafluorophenyl)dimethylsilane ≥97.0%
Bis(pentafluorophenyl)dimethylsilane

CAS Number: 10536-62-6
MDL Number: MFCD00042066
Molecular Formula: C14H6F10Si
Molecular Weight: 392.27
Purity/Analysis Method: 97.0% (GC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Bis(perfluorophenyl)dimethylsilane

Formula: C₁₄H₆F₁₀Si
MDL Number: MFCD00042066
CAS Number: 10536-62-6
https://us.vwr.com/stibo/web/std.lang.all/40/40/28934040.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR