1-Butyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B2243-5G
B2243-25G
TCB2243-25GEA
486
USD
TCB2243-25G
76219-352
1-Butyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)
1-Butyl-1-phenylhydrazine
CAS Number: 61715-75-1
MDL Number: MFCD03844777
Molecular Formula: C10H16N2
Molecular Weight: 164.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 105
Specific Gravity (20/20): 0.98
Formula:
C₁₀H₁₆N₂ Boiling Pt: 105 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD03844777 CAS Number: 61715-75-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...