2-Arsanilic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: o-Arsanilic acid2-Aminophenylarsonic acid2-Aminobenzenearsonic acid

A0529-25G A0529-5G
TCA0529-025GEA 103.48 USD
TCA0529-025G TCA0529-005G
2-Arsanilic acid ≥98.0% (by titrimetric analysis)
2-Arsanilic acid

CAS Number: 2045-00-3
MDL Number: MFCD00007630
Molecular Formula: C6H8AsNO3
Molecular Weight: 217.06
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 153


Other Product Names / Synonyms: 2-Aminobenzenearsonic Acid, o-Arsanilic Acid

Formula: C₆H₈AsNO₃
MW: 217.06 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00007630
CAS Number: 2045-00-3
EINECS: 218-064-5
UN: 3465
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/79/59/28927959.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR