5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate ≥94.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Cyclohexyl N-Boc-L-glutamateBoc-Glu(OcHex)-OHN-Boc-L-glutamic Acid 5-Cyclohexyl EsterN-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Cyclohexyl Ester

B2179-5G
TCB2179-5GEA 51 USD
TCB2179-5G
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate ≥94.0% (by titrimetric analysis)
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate

CAS Number: 73821-97-3
MDL Number: MFCD00065570
Molecular Formula: C16H27NO6
Molecular Weight: 329.39
Purity/Analysis Method: 94.0% (T)
Form: Crystal
Melting point (°C): 57
Specific rotation [a]20/D: -16 deg (C=2, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-glutamic Acid 5-Cyclohexyl Ester, N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Cyclohexyl Ester, 5-Cyclohexyl N-Boc-L-glutamate

Formula: C₁₆H₂₇NO₆
MW: 329.39 g/mol
Melting Pt: 57 °C
MDL Number: MFCD00065570
CAS Number: 73821-97-3
https://us.vwr.com/stibo/web/std.lang.all/39/48/28933948.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR