Bis(phenylsulfonyl)methane ≥98.0%
Supplier: TCI America
B2167-5G
TCB2167-5GEA
132.97
USD
TCB2167-5G
Bis(phenylsulfonyl)methane ≥98.0%
Bis(phenylsulfonyl)methane
CAS Number: 3406-02-8
MDL Number: MFCD00007553
Molecular Formula: C13H12O4S2
Molecular Weight: 296.36
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 121
Formula:
C₁₃H₁₂O₄S₂ Melting Pt: 121 °C |
MDL Number:
MFCD00007553 CAS Number: 3406-02-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...