3-Fluorophenethyl bromide ≥97.0%
Supplier: TCI America
Synonyms:
1-(2-Bromoethyl)-3-fluorobenzene
B2159-5G
TCB2159-5GEA
339.92
USD
TCB2159-5G
3-Fluorophenethyl bromide ≥97.0%
3-Fluorophenethyl bromide
CAS Number: 25017-13-4
MDL Number: MFCD03095397
Molecular Formula: C8H8BrF
Molecular Weight: 203.05
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 94
Specific Gravity (20/20): 1.45
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(2-Bromoethyl)-3-fluorobenzene, 3-Fluorophenethyl Bromide
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...