TCIAB2153

B2153-ZONE
TCB2153-ZONEEA 794.21 USD
TCB2153-ZONE
TCIAB2153
CAS Number: 1625-92-9
MDL Number: MFCD00222366
Molecular Formula: C16H18
Molecular Weight: 210.32
Boiling point (°C): 310
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR