DL-Arabitol ≥97.0%
Supplier: TCI America
A0517-100MG
TCA0517-100MGEA
32.32
USD
TCA0517-100MG
DL-Arabitol ≥97.0%
DL-Arabitol
CAS Number: 2152-56-9
MDL Number: MFCD00070503
Molecular Formula: C5H12O5
Molecular Weight: 152.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 106
Product Categories: Bioscience, Sugar Alcohols, Arabinoses
Other Product Names / Synonyms: DL-Arabinitol
Formula:
C₅H₁₂O₅ Melting Pt: 106 °C |
MDL Number:
MFCD00070503 CAS Number: 6018-27-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...