(R)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane ≥97.0%
Supplier: TCI America
B2136-1G
TCB2136-001GEA
155
USD
TCB2136-001G
(R)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane ≥97.0%
(R)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane
CAS Number: 14347-83-2
MDL Number: MFCD00066533
Molecular Formula: C13H18O3
Molecular Weight: 222.28
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: -19 deg (neat)
Formula:
C₁₃H₁₈O₃ Density: 1.05 (20 °C) |
MDL Number:
MFCD00066533 CAS Number: 14347-83-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...