2-Bromo-1,3,2-benzodioxaborole (β-bromocatecholborane) ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Bromo-1,3,2-benzodioxaborole
, B-Bromocatecholborane
B2132-25G
B2132-5G
TCB2132-005GEA
98
USD
TCB2132-005G
TCB2132-25G
2-Bromo-1,3,2-benzodioxaborole (β-bromocatecholborane) ≥97.0% (by titrimetric analysis)
2-Bromo-1,3,2-benzodioxaborole (β-Bromocatecholborane)
CAS Number: 51901-85-0
MDL Number: MFCD00013341
Molecular Formula: C6H4BBrO2
Molecular Weight: 198.81
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 76
Melting point (°C): 52
Flash Point (°C): 42
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Bromo-1,3,2-benzodioxaborole
Formula:
C₆H₄BBrO₂ Boiling Pt: 76 °C Melting Pt: 52 °C Flash Pt: 42 °C |
MDL Number:
MFCD00013341 CAS Number: 51901-85-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...