(S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane bis(borane)
Synonyms:
Borane - (S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane Complex
, (S,S)-1,2-Bis[boranato(tert-butyl)methylphosphino]ethane
B2089-100MG
TCB2089-100MGEA
182.04
USD
TCB2089-100MG
(S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane bis(borane)
(S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane bis(borane)
CAS Number: 203000-48-0
MDL Number: MFCD03844770
Molecular Formula: C12H34B2P2
Molecular Weight: 261.97
Form: Crystal
Color: White
Other Product Names / Synonyms: (S,S)-1,2-Bis[boranato(tert-butyl)methylphosphino]ethane, Borane - (S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane Complex
Formula:
C₁₂H₃₄B₂P₂ MW: 261.97 g/mol |
MDL Number:
MFCD03844770 CAS Number: 203000-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...