Methyl-3,5-bis(tert-butyldiphenylsilyloxy)benzoate ∼20% ∼0.28 mol/L in toluene
Supplier: TCI America
Synonyms:
3,5-Bis(tert-butyldiphenylsilyloxy)benzoic Acid Methyl Ester
B2051-25G
TCB2051-025GEA
1313
USD
TCB2051-025G
Methyl-3,5-bis(tert-butyldiphenylsilyloxy)benzoate ∼20% ∼0.28 mol/L in toluene
Methyl-3,5-bis(tert-butyldiphenylsilyloxy)benzoate
CAS Number: 182250-68-6
MDL Number: MFCD02093439
Molecular Formula: C40H44O4Si2
Molecular Weight: 644.96
Form: Clear Liquid
Color: Colorless
Other Product Names / Synonyms: 3,5-Bis(tert-butyldiphenylsilyloxy)benzoic Acid Methyl Ester
Formula:
C₄₀H₄₄O₄Si₂ MW: 644.96 g/mol |
MDL Number:
MFCD02093439 CAS Number: 182250-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...