Bis(methylthio)(trimethylsilyl)methane ≥98.0%

Supplier: TCI America

Synonyms: Bis(methylthio)methyltrimethylsilaneTrimethylbis(methylthio)methylsilane

B2004-1G
TCB2004-001GEA 110 USD
TCB2004-001G
Bis(methylthio)(trimethylsilyl)methane ≥98.0%
Bis(methylthio)(trimethylsilyl)methane

CAS Number: 37891-79-5
MDL Number: MFCD01631301
Molecular Formula: C6H16S2Si
Molecular Weight: 180.40
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid


Product Categories: Synthetic Organic Chemistry, Sulfides, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Bis(methylthio)methyltrimethylsilane, Trimethylbis(methylthio)methylsilane

Formula: C₆H₁₆S₂Si
MW: 180.40 g/mol
MDL Number: MFCD01631301
CAS Number: 37891-79-5
https://us.vwr.com/stibo/web/std.lang.all/37/62/28933762.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR