(S)-(-)-3-tert-Butylamino-1,2-propanediol ≥96.0%

Supplier: TCI America

Synonyms: (S)-3-(tert-Butylamino)propane-1,2-diol

B1987-5G
TCB1987-005GEA 123 USD
TCB1987-005G
(S)-(-)-3-tert-Butylamino-1,2-propanediol ≥96.0%
(S)-(-)-3-tert-Butylamino-1,2-propanediol

CAS Number: 30315-46-9
MDL Number: MFCD00190165
Molecular Formula: C7H17NO2
Molecular Weight: 147.22
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Melting point (°C): 81
Specific rotation [a]20/D: -29 deg (C=2, 1mol/L HCl)

Formula: C₇H₁₇NO₂
MW: 147.22 g/mol
MDL Number: MFCD00190165
CAS Number: 30315-46-9

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR