2-tert-Butyl-1,4-benzoquinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1984-25G
TCB1984-025GEA 102.26 USD
TCB1984-025G
2-tert-Butyl-1,4-benzoquinone ≥98.0% (by GC, titration analysis)
2-tert-Butyl-1,4-benzoquinone

CAS Number: 3602-55-9
MDL Number: MFCD00666928
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 91

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Melting Pt: 57 °C
Flash Pt: 91 °C
MDL Number: MFCD00666928
CAS Number: 3602-55-9
https://us.vwr.com/stibo/web/std.lang.all/37/45/28933745.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR