Bis(tributylstannyl)acetylene ≥95.0%

B1974-5G
TCB1974-005GEA 269.73 USD
TCB1974-005G
Bis(tributylstannyl)acetylene ≥95.0%
Bis(tributylstannyl)acetylene

CAS Number: 994-71-8
MDL Number: MFCD00010239
Molecular Formula: C26H54Sn2
Molecular Weight: 604.14
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110

Formula: C₂₆H₅₄Sn₂
Flash Pt: 110 °C
MDL Number: MFCD00010239
CAS Number: 994-71-8

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR