o-Anisidinium chloride ≥99.0% (by HPLC)
Supplier: TCI America
Synonyms:
2-Methoxyaniline hydrochloride
A0489-25G
TCA0489-025GEA
133.92
USD
TCA0489-025G
o-Anisidinium chloride ≥99.0% (by HPLC)
o-Anisidinium chloride
CAS Number: 134-29-2
MDL Number: MFCD00054334
Molecular Formula: C7H9NO
Molecular Weight: 159.61
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2-Methoxyaniline Hydrochloride
Formula:
C₇H₁₀ClNO MW: 159.6134 g/mol |
MDL Number:
MFCD00054334 CAS Number: 134-29-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...