m-Anisidinium chloride ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: m-Anisidine hydrochloride3-Methoxyaniline hydrochloride

A0488-25G
TCA0488-025GEA 142.04 USD
TCA0488-025G
m-Anisidinium chloride ≥99.0% (by HPLC, total nitrogen)
m-Anisidinium chloride

CAS Number: 27191-09-9
MDL Number: MFCD00060220
Molecular Formula: C7H9NO
Molecular Weight: 159.61
Purity/Analysis Method: 99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 174


Other Product Names / Synonyms: 3-Methoxyaniline Hydrochloride

Formula: C₇H₁₀ClNO
MW: 159.6100 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00060220
CAS Number: 27191-09-9
https://us.vwr.com/stibo/web/std.lang.all/79/21/28927921.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR