1H-Benzotriazole-1-methanol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(Hydroxymethyl)benzotriazole

B1903-25G
TCB1903-025GEA 52.78 USD
TCB1903-025G
1H-Benzotriazole-1-methanol ≥98.0% (by titrimetric analysis)
1H-Benzotriazole-1-methanol

CAS Number: 28539-02-8
MDL Number: MFCD00179118
Molecular Formula: C7H7N3O
Molecular Weight: 149.15
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 150


Other Product Names / Synonyms: 1-(Hydroxymethyl)-1H-benzotriazole

Formula: C₇H₇N₃O
MW: 149.1500 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00179118
CAS Number: 28539-02-8
https://us.vwr.com/stibo/web/std.lang.all/36/72/28933672.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR