(1S,2S)-N,N'-Bis[3-oxo-2-(2,4,6-trimethylbenzoyl)butylidene]-1,2-diphenylethylenediaminato cobalt(II)
Synonyms:
N,N'-Bis[2-(mesitoyl)-3-oxobutylidene]-(1S,2S)-1,2-diphenylethylenediaminato Cobalt(II)
B1845-100MG
TCB1845-100MGEA
72.42
USD
TCB1845-100MG
(1S,2S)-N,N'-Bis[3-oxo-2-(2,4,6-trimethylbenzoyl)butylidene]-1,2-diphenylethylenediaminato cobalt(II)
(1S,2S)-N,N'-Bis[3-oxo-2-(2,4,6-trimethylbenzoyl)butylidene]-1,2-diphenylethylenediaminato cobalt(II)
CAS Number: 171200-71-8
MDL Number: MFCD01321132
Molecular Formula: C42H42CoN2O4
Molecular Weight: 697.74
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: N,N'-Bis[2-(mesitoyl)-3-oxobutylidene]-(1S,2S)-1,2-diphenylethylenediaminato Cobalt(II), (S)-MPAC
Formula:
C₄₂H₄₂CoN₂O₄ MW: 697.74 g/mol |
MDL Number:
MFCD01321132 CAS Number: 171200-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...