TCIAB1844

B1844-100MG
TCB1844-100MGEA 370.61 USD
TCB1844-100MG
TCIAB1844
CAS Number: 212250-92-5
MDL Number: MFCD01321131
Molecular Formula: C42H42CoN2O4
Molecular Weight: 697.74
Purity/Analysis Method: 90.0% (T)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Organometallic Catalysts
Other Product Names / Synonyms: N,N'-Bis[2-(mesitoyl)-3-oxobutylidene]-(1R,2R)-1,2-diphenylethylenediaminato Cobalt(II), (R)-MPAC
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR