Bis(hexamethylene)triamine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bis(6-aminohexyl)amine6,6'-Iminodihexylamine

B1814-400G B1814-25G
TCB1814-025GEA 30.43 USD
TCB1814-025G TCB1814-400G
Bis(hexamethylene)triamine ≥95.0% (by titrimetric analysis)
Bis(hexamethylene)triamine

CAS Number: 143-23-7
MDL Number: MFCD00010448
Molecular Formula: C12H29N3
Molecular Weight: 215.39
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 35
Flash Point (°C): 121


Product Categories: Bioscience, Polyamines
Other Product Names / Synonyms: Bis(6-aminohexyl)amine, 6,6'-Iminodihexylamine

Formula: C₁₂H₂₉N₃
MW: 215.39 g/mol
Boiling Pt: 164 °C
Melting Pt: 35 °C
Flash Pt: 121 °C
MDL Number: MFCD00010448
CAS Number: 143-23-7
https://us.vwr.com/stibo/web/std.lang.all/35/88/28933588.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR