1,4-Di(chloromethyl)-2,3,5,6-tetramethylbenzene ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
3,6-Bis(chloromethyl)durene
, 1,4-Bis(chloromethyl)-2,3,5,6-tetramethylbenzene
B1783-5G
TCB1783-005GEA
140.01
USD
TCB1783-005G
1,4-Di(chloromethyl)-2,3,5,6-tetramethylbenzene ≥98.0%
1,4-Di(chloromethyl)-2,3,5,6-tetramethylbenzene
CAS Number: 3022-16-0
MDL Number: MFCD00018883
Molecular Formula: C12H16Cl2
Molecular Weight: 231.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 196
Other Product Names / Synonyms: 1,4-Bis(chloromethyl)-2,3,5,6-tetramethylbenzene
Formula:
C₁₂H₁₆Cl₂ MW: 231.16 g/mol Melting Pt: 196 °C |
MDL Number:
MFCD00018883 CAS Number: 3022-16-0 EINECS: 221-173-0 UN: 3335 ADR: 9, |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...