8-Anilino-1-naphthalenesulfonic acid ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 8-(Phenylamino)naphthalene-1-sulphonic acid8-Anilinonaphthalene-1-sulfonic acid8-Anilinonaphthalene-1-sulphonic acid8-(Phenylamino)naphthalene-1-sulfonic acid

A0472-25G
TCA0472-025GEA 86.24 USD
TCA0472-025G
8-Anilino-1-naphthalenesulfonic acid ≥95.0% (by HPLC, titration analysis)
8-Anilino-1-naphthalenesulphonic acid

CAS Number: 82-76-8
MDL Number: MFCD00003998
Molecular Formula: C16H13NO3S
Molecular Weight: 299.34
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 217


Product Categories: Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Phenyl Acid, N-Phenyl-1-naphthylamine-8-sulfonic Acid, Phenyl Peri Acid

Formula: C₁₆H₁₃NO₃S
MW: 299.35 g/mol
Melting Pt: 217 °C
MDL Number: MFCD00003998
CAS Number: 82-76-8
EINECS: 201-438-7
https://us.vwr.com/stibo/web/std.lang.all/79/03/28927903.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR