Methyl docosanoate ≥96.0%

Supplier: TCI America

Synonyms: Behenic acid methyl ester Docosanoic acid methyl ester Methyl behenate

B1748-10G
TCB1748-010GEA 183.7 USD
TCB1748-010G
Methyl docosanoate ≥96.0%
Methyl docosanoate

CAS Number: 929-77-1
MDL Number: MFCD00009347
Molecular Formula: C23H46O2
Molecular Weight: 354.62
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Boiling point (°C): 393
Melting point (°C): 55


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Behenic Acid Methyl Ester, Docosanoic Acid Methyl Ester, Methyl Docosanoate

Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
Boiling Pt: 393 °C
Melting Pt: 55 °C
MDL Number: MFCD00009347
CAS Number: 929-77-1
EINECS: 213-207-8
https://us.vwr.com/stibo/web/std.lang.all/34/93/28933493.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR